Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human OTOR Recombinant Protein, PeproTech®

Catalog No. 13003100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13003100UG 100 μg
130-03-1MG 1 mg
130-03-20UG 20 μg
13003250UG 250 μg
13003500UG 500 μg
130-03-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 13003100UG Supplier Gibco™ Supplier No. 13003100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human OTOR is a 12.7 kDa globular protein containing 112 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

OTOR, also called Otoraplin and MIAL, is a secreted cytokine of the MIA/OTOR family. Members of this family, which include MIA, MIA2, and TANGO, share a Src homology-3 (SH3)-like domain. OTOR is predominantly expressed in the cochlea of the inner ear, and, to a lesser extent, in fetal brain tissue and some cartilage tissues. OTOR appears to be involved in early chondrogenesis of the otic capsule, which is required for normal inner ear development and auditory function.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NRC9
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 56914
Molecular Weight (g/mol) 12.7 kDa
Name Human OTOR
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CDRAP; FDP; Fibrocyte-derived protein; melanoma inhibitory activity-like protein; MIA; MIAL; MIAL1; Otor; Otoraplin; UNQ3054/PRO9873
Common Name OTOR
Gene Symbol OTOR
Conjugate Unconjugated
Recombinant Recombinant
Sequence MVHGIFMDRL ASKKLCADDE CVYTISLASA QEDYNAPDCR FINVKKGQQI YVYSKLVKENGAGEFWAGSV YGDGQDEMGV VGYFPRNLVK EQRVYQEATK EVPTTDIDFF CE
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.