Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human OTP Full-length ORF (NP_115485.1, 1 a.a. - 325 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89955397 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-955-397 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-955-397 Supplier Abnova™ Supplier No. H00023440P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the homeodomain (HD) family. HD family proteins are helix-turn-helix transcription factors that play key roles in the specification of cell fates. This protein may function during brain development. [provided by RefSeq]

Sequence: MLSHADLLDARLGMKDAAELLGHREAVKCRLGVGGSDPGGHPGDLAPNSDPVEGATLLPGEDITTVGSTPASLAVSAKDPDKQPGPQGGPNPSQAGQQQGQQKQKRHRTRFTPAQLNELERSFAKTHYPDIFMREELALRIGLTESRVQVWFQNRRAKWKKRKKTTNVFRAPGTLLPTPGLPQFPSAAAAAAAAMGDSLCSFHANDTRWAAAAMPGVSQLPLPPALGRQQAMAQSLSQCSLAAGPPPNSMGLSNSLAGSNGAGLQSHLYQPAFPGMVPASLPGPSNVSGSPQLCSSPDSSDVWRGTSIASLRRKALEHTVSMSFT

Specifications

Accession Number NP_115485.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23440
Molecular Weight (g/mol) 60.6kDa
Name OTP (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MLSHADLLDARLGMKDAAELLGHREAVKCRLGVGGSDPGGHPGDLAPNSDPVEGATLLPGEDITTVGSTPASLAVSAKDPDKQPGPQGGPNPSQAGQQQGQQKQKRHRTRFTPAQLNELERSFAKTHYPDIFMREELALRIGLTESRVQVWFQNRRAKWKKRKKTTNVFRAPGTLLPTPGLPQFPSAAAAAAAAMGDSLCSFHANDTRWAAAAMPGVSQLPLPPALGRQQAMAQSLSQCSLAAGPPPNSMGLSNSLAGSNGAGLQSHLYQPAFPGMVPASLPGPSNVSGSPQLCSSPDSSDVWRGTSIASLRRKALEHTVSMSFT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC3161
Common Name OTP
Gene Symbol OTP
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.