Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human OVGP1 Partial ORF (NP_002548.3, 579 a.a. - 677 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89955420 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-955-420 25 ug
89-955-419 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-955-420 Supplier Abnova™ Supplier No. H00005016Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a large, carbohydrate-rich, epithelial glycoprotein with numerous O-glycosylation sites located within threonine, serine, and proline-rich tandem repeats. The gene is similar to members of the mucin and the glycosyl hydrolase 18 gene families. Regulation of expression may be estrogen-dependent. Gene expression and protein secretion occur during late follicular development through early cleavage-stage embryonic development. The protein is secreted from non-ciliated oviductal epithelial cells and associates with ovulated oocytes, blastomeres, and spermatozoan acrosomal regions. [provided by RefSeq]

Sequence: RNISVTPEGQTMPLRGENLTSEVGTHPRMGNLGLQMEAENRMMLSSSPVIQLPEQTPLAFDNRFVPIYGNHSSVNSVTPQTSPLSLKKEIPENSAVDEE

Specifications

Accession Number NP_002548.3
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5016
Molecular Weight (g/mol) 36.63kDa
Name OVGP1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen RNISVTPEGQTMPLRGENLTSEVGTHPRMGNLGLQMEAENRMMLSSSPVIQLPEQTPLAFDNRFVPIYGNHSSVNSVTPQTSPLSLKKEIPENSAVDEE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CHIT5/EGP/MUC9/OGP
Common Name OVGP1
Gene Symbol OVGP1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.