Learn More
R&D Systems™ Human OX40/TNFRSF4 DuoSet ELISA
Description
Specifications
Specifications
| Assay Range | 31.20 to 2000 pg/mL |
| Product Type | Solid Phase Sandwich ELISA |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7293 |
| Quantity | 1 Kit (for 5 Plates) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | ACT-135, ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, Ly-70, O x 40 cell surface antigen, O x 40 homologue, O x 40 L receptor, O x 40 lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, TNFRSF4, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, Txgp1, TXGP1L, TXGP1LO x 40 antigen |
| Target | OX40/TNFRSF4 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.