Learn More
ILEX LIFE SCIENCES LLC HUMAN OXYTOCIN SYNTHETIC 100MG
SDP
Supplier:  ILEX LIFE SCIENCES LLC A2254100
Encompass_Preferred
Ilex Life Sciences Oxytocin (OT) Human, Synthetic Protein Hormone (100 mg). DESCRIPTION: Our Human Oxytocin (OT) synthetic protein hormone is a single, non-glycosylated, polypeptide chain containing 9 amino acids and has a molecular mass of 1007.2 Dalton. Oxytocin has a molecular formula of C43H66N12O12S2. CAS REGISTRY NO.: 50-56-6. PURITY: Greater than 98.0% as determined by RP-HPLC. AMINO ACID SEQUENCE: H-Cys-Tyr-Ile-Gln-Asn-Cys-Pro-Leu-Gly-NH2. FORMULATION: Sterile filtered white lyophilized powder. The Oxytocin was lyophilized from a concentrated (1 mg/ml) solution with no additives. SYNONYMS: OT, Oxytocin-neurophysin 1, OT-NPI, Ocytocin, Neurophysin 1, α-Hypophamine, Oxytocic hormone, OXT gene product. USAGE: For laboratory research use only; not for use in diagnostic or therapeutic procedures.
Catalog No. 50-273-4008
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.