Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human P-selectin (CD62P) Fc Chimera Recombinant Protein

Catalog No. PIRP88162
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88162 100 μg
PIRP88163 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP88162 Supplier Invitrogen™ Supplier No. RP88162
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (400 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.

P-selectin (Selectin P, GMP-140, LECAM3, CD62 antigen-like family member) is a 140 kDa type 1 transmembrane glycoprotein, and is one of the most commonly studied proteins that regulate cell rolling. P-selectin is stored in the Weibel-Palade bodies of endothelial cells, as well as in a-granules of platelets. From there, it can be rapidly brought to the cell surface after exposure to thrombin, histamine, complement 5a, Ca21 ionophores, or adenosine diphosphate. P-selectin protein redistributes to the plasma membrane during platelet activation and degranulation and mediates the interaction of activated endothelial cells or platelets with leukocytes. P-selectin plays an important role in adhesive processes between leucocytes and endothelial cells, and is a calcium dependent receptor that binds to sialylated forms of Lewis blood group carbohydrate antigens found on neutrophils and monocytes. P-selectin is constitutively expressed in inflammation and contributes to atherogenesis, thrombosis and tissue destruction. Clinically, P-selectin is used to distinguish heparin induced thrombocytopenia with and without thrombosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P16109
Concentration 0.25 mg/mL
For Use With (Application) Control, Bioactivity
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 6403
Molecular Weight (g/mol) 106.97 kDa
Name Human P-selectin (CD62P) Fc Chimera
pH Range 7.4
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias CD62; CD62 antigen-like family member P; CD62P; CD62P antigen; CD62P; CD62 homolog; FLJ45155; GMP140; GMP-140; GMRP; Granule membrane protein 140; granule membrane protein 140kDa; granulocyte membrane protein; Grmp; LECAM3; leukocyte-endothelial cell adhesion molecule 3; PADGEM; Platelet activation dependent granule-external membrane protein; platelet alpha-granule membrane protein; PSEL; PSELECT; P-selectin; P-selectin precursor; RP1-86F14.2; selectin P; selectin P (granule membrane protein 140kDa, antigen CD62); selectin, platelet; Selp
Common Name P-Selectin (CD62P)
Gene Symbol SELP
Biological Activity Measured by the ability of the immobilized protein to support the adhesion of U937 human histiocytic lymphoma cells. When 5 x 10E4 cells/well are added to SELP/Fc Chimera coated plates (10 μg/mL with 100 μL/well), > 80% cells will adhere after 1 hour (at 37°C.)
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Human P-selectin, amino acids Met1-Ala771 (Accession # AAN06828.1) with a C-terminal human IgG1 Fc region
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Measured by the ability of the immobilized protein to support the adhesion of U937 human histiocytic lymphoma cells. When 5 x 10E4 cells/well are added to SELP/Fc Chimera coated plates (10 μg/mL with 100 μL/well), > 80% cells will adhere after 1 hour at 37°C.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥85% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.