Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human P-Selectin/CD62P Quantikine ELISA Kit
SDP

Catalog No. DPSE00 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DPSE00 1 Kit
SPSE00 1 Pack (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DPSE00 Supplier R&D Systems™ Supplier No. DPSE00

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human P-Selectin/CD62P Quantikine ELISA Kit quantifies human P-Selectin/CD62P in cell culture supernates (10 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), citrate plasma (10 ul).

1 Kit

Specifications

Assay Range 0.78 - 50 ng/mL
Assay Sensitivity 0.121 ng/mL
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human P-Selectin
For Use With (Application) ELISA
Gene ID (Entrez) 6403
Host Species Human
Quantity 1 Kit
Sample Volume Cell Culture Supernates (10 uL),Serum (10 uL),EDTA Plasma (10 uL),Heparin Plasma (10 uL),Citrate Plasma (10 uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CD62P, CD62P antigen, FLJ45155, GMP140, GRMP, PADGEM, PADGEMantigen CD62), PSEL, selectin P (granule membrane protein 140 kDa, antigen CD62), SELP
Target P-Selectin/CD62P
Total Assay Time 1.25 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.