Learn More
Gibco™ Human p16-INK4a Recombinant Protein, PeproTech®
Description
110-02-1MG will be provided as 10 x 100 μg (110-02-100UG). 110-02-500UG will be provided as 5 x 100 μg (110-02-100UG). Recombinant Human p16-INK4a is a 16.5 kDa protein containing 156 amino acid residues. Kim, W.Y. and Sharpless, N.E. (2006)Cell 127(2):265-75. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P42771 |
| For Use With (Application) | Control |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 1029 |
| Molecular Weight (g/mol) | 16.5 kDa |
| Name | Human p16-INK4a |
| Purification Method | Purified |
| Quantity | 5 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.