Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human p16-INK4a-TAT Recombinant Protein, PeproTech®

Catalog No. 11002T500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
5 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11002T500UG 500 μg
11002T100UG 100 μg
110-02T-1MG 1 mg
11002T25UG 25 μg
110-02T-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 11002T500UG Supplier Gibco™ Supplier No. 11002T500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human p16-INK4a-TAT expressed in E.coli is an 18.0 kDa protein containing 167 amino-acid residues, including the 155 residues of full-length p16-INK4a and a 12-residue C-terminal TAT peptide (GYGRKKRRQRRR). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

p16-INK4a is a nuclear protein that regulates the cell cycle by inhibiting cyclin-dependent kinase-4 (CDK4) and CDK6. p16-INK4a inhibits CDK activity by binding to the CDK molecules in a manner that interferes with their ability to interact with cyclin D. This activity has the effect of suppressing tumor formation and growth, and of inducing replicative senescence in various normal cells, including stem cells. The expression of p16-INK4a steadily increases with age, and tends to accumulate in stem cell compartments. The deletion, rearrangement, or mutation of the p16-INK4a gene is frequently found in melanomas, as well as in certain other types of cancer. p16-INK4a and other transcription factors have been introduced into cells by DNA transfection, viral infection, or microinjection. Protein transduction using TAT fusion proteins represents an alternative methodology for introducing transcription factors and other nuclear proteins into primary, as well as transformed, cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P42771, Q8N726
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 1029
Molecular Weight (g/mol) 18 kDa
Name Human p16-INK4a-TAT
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias p16INK4a
Common Name p16INK4a-TAT
Gene Symbol CDKN2A
Conjugate Unconjugated
Recombinant Recombinant
Sequence EPAAGSSMEP SADWLATAAA RGRVEEVRAL LEAGALPNAP NSYGRRPIQV MMMGSARVAE LLLLHGAEPN CADPATLTRP VHDAAREGFL DTLVVLHRAG ARLDVRDAWG RLPVDLAEEL GHRDVARYLR AAAGGTRGSN HARIDAAEGP SDIPDGYGR KKRRQRRR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.