Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PA2G4 Full-length ORF (NP_006182.2, 1 a.a. - 394 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89955526 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-955-526 25 ug
89-955-525 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-955-526 Supplier Abnova™ Supplier No. H00005036P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes an RNA-binding protein that is involved in growth regulation. This protein is present in pre-ribosomal ribonucleoprotein complexes and may be involved in ribosome assembly and the regulation of intermediate and late steps of rRNA processing. This protein can interact with the cytoplasmic domain of the ErbB3 receptor and may contribute to transducing growth regulatory signals. This protein is also a transcriptional co-repressor of androgen receptor-regulated genes and other cell cycle regulatory genes through its interactions with histone deacetylases. This protein has been implicated in growth inhibition and the induction of differentiation of human cancer cells. Six pseudogenes, located on chromosomes 3, 6, 9, 18, 20 and X, have been identified. [provided by RefSeq]

Sequence: MSGEDEQQEQTIAEDLVVTKYKMGGDIANRVLRSLVEASSSGVSVLSLCEKGDAMIMEETGKIFKKEKEMKKGIAFPTSISVNNCVCHFSPLKSDQDYILKEGDLVKIDLGVHVDGFIANVAHTFVVDVAQGTQVTGRKADVIKAAHLCAEAALRLVKPGNQNTQVTEAWNKVAHSFNCTPIEGMLSHQLKQHVIDGEKTIIQNPTDQQKKDHEKAEFEVHEVYAVDVLVSSGEGKAKDAGQRTTIYKRDPSKQYGLKMKTSRAFFSEVERRFDAMPFTLRAFEDEKKARMGVVECAKHELLQPFNVLYEKEGEFVAQFKFTVLLMPNGPMRITSGPFEPDLYKSEMEVQDAELKALLQSSASRKTQKKKKKKASKTAENATSGETLEENEAGD

Specifications

Accession Number NP_006182.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5036
Molecular Weight (g/mol) 70.2kDa
Name PA2G4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MSGEDEQQEQTIAEDLVVTKYKMGGDIANRVLRSLVEASSSGVSVLSLCEKGDAMIMEETGKIFKKEKEMKKGIAFPTSISVNNCVCHFSPLKSDQDYILKEGDLVKIDLGVHVDGFIANVAHTFVVDVAQGTQVTGRKADVIKAAHLCAEAALRLVKPGNQNTQVTEAWNKVAHSFNCTPIEGMLSHQLKQHVIDGEKTIIQNPTDQQKKDHEKAEFEVHEVYAVDVLVSSGEGKAKDAGQRTTIYKRDPSKQYGLKMKTSRAFFSEVERRFDAMPFTLRAFEDEKKARMGVVECAKHELLQPFNVLYEKEGEFVAQFKFTVLLMPNGPMRITSGPFEPDLYKSEMEVQDAELKALLQSSASRKTQKKKKKKASKTAENATSGETLEENEAGD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias EBP1/HG4-1/p38-2G4
Common Name PA2G4
Gene Symbol PA2G4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.