Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PAI-1 (Serpin E1) Recombinant Protein

Catalog No. PIRP75687
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP75687 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP75687 Supplier Invitrogen™ Supplier No. RP75687
Add to Cart
Add to Cart

Recombinant Protein

This product contains purified recombinant human Plasminogen Activator Inhibitor 1 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

PAI1 (plasminogen activator inhibitor 1) belongs to serine protease inhibitor superfamily, and is the principal inhibitor of tissue-type and urokinase-type plasminogen activators (tPA and uPA). Platelets are the main source of the circulating PAI11, but it is synthesized and secreted by many tissue and cell types, including fibroblasts, smooth muscle cells, endothelial cells, hepatocytes, and inflammatory cells. Expression of PAI11 can be regulated at the transcriptional level by many factors including growth factors, cytokines, hormones, inflammatory factors, glucose or lipid metabolites, vascular tone regulating factors, chemicals, and other environmental or physical factors. PAI1 is present at increased levels in various disease states, and has been linked to an increased occurrence of thrombosis in obesity, thrombophilia and the metabolic syndrome. Defects in PAI-1 are characterized by abnormal bleeding. PAI1 mediates inhibition of fibrinolysis by inhibiting the activity of plasminogen activator, and may promote neuronal survival. Other defects in PAI1 are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency). Alternatively spliced transcript variants encoding different isoforms of PAI1 have been found.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05121
Concentration 0.10 mg/mL
For Use With (Application) Control, Western Blot
Formulation 0.025 M sodium acetate with 50% glycerol and no preservative; pH 4.8
Gene ID (Entrez) 5054
Name Human PAI-1 (Serpin E1)
pH Range 4.8
Quantity 1 mg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias Endothelial plasminogen activator inhibitor; nexin, plasminogen activator inhibitor type 1; PAI; PAI1; PAI-1; PAI1 protein; PAI1A; Pai1aa; Planh; Planh1; plas; plasminogen activator inhibitor; plasminogen activator inhibitor 1; plasminogen activator inhibitor I; plasminogen activator inhibitor, type I; plasminogen activator inhibitor-1; Plasminogen activator inhibitor-1 precursor (PAI-1) (Endothelial plasminogen activator inhibitor) (PAI); putative plasminogen activator inhibitor-1; RATPAI1A; serine (or cysteine) peptidase inhibitor, clade E, member 1; serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1; serine (or cysteine) proteinase inhibitor, clade E, member 1; serine (or cysteine) proteinase inhibitor, member 1; serpin E1; serpin family E member 1; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1; SERPINE1; SERPINE-1
Common Name PAI1
Gene Symbol SERPINE1
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Val24-Pro402 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.