Learn More
Cellular Engineering Technologies Cryopreserved vial of Panc-1 carcinoma cells(500,000 cells)
Supplier:  Cellular Engineering Technologies CR1016500
Encompass_Preferred
Human pancreatic carcinoma cells with ductal origin display an epithelial morphology. These cells are from a 56-year-old Caucasian male donor. This cell line grows adherently and displays robust growth characteristics.These cell types are routinely used to study the role of the intermediate filament keratin reorganization in the migration of cancer cells. These Panc-1 cells are homogeneous and grow robustly in cell culture. Vial contains approximately 500,000 cells. Shipped with dry ice.
Catalog No. NC2753056
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.