Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Pancreatic Polypeptide/PP ELISA Kit

Catalog No. EH357RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH357RBX5 5 x 96 Tests
EH357RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH357RBX5 Supplier Invitrogen™ Supplier No. EH357RBX5
Add to Cart
Add to Cart

ELISA

Human Pancreatic Polypeptide/PP quantitates human Pancreatic Polypeptide/PP in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Pancreatic Polypeptide/PP.

Pancreatic polypeptide (PP) is a 36 amino-acid residue peptide with sequence and 3-D structural homology with NPY and PYY. PP is the least conserved of the three peptides and was the first to be identified. It is almost exclusively expressed in the endocrine pancreas and released in response to cholinergic stimulation and food intake. PP has inhibitory effects on gall bladder secretion, intestinal motility and pancreatic secretion. PP may serve as a marker for certain gut endocrine tumors including carcinoid tumors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AV061413; C-terminal peptide; HGNC:9327; IGSPP; IOPPP; Obinepitide; panceatic polypeptide; Pancreatic hormone; Pancreatic icosapeptide; Pancreatic polypeptide; pancreatic polypeptide Y; pancreatic prohormone; Pancreatic prohormone percursor; pancreatic prohormone precursor; PH; PI; PNP; PP; PP1; PPA1; PPase; Ppy; prepro-PP (prepropancreatic polypeptide); SID6-8061
Assay Range 4 to 1,000 pg/mL
Assay Sensitivity 4 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5539
Gene Symbol PPY
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target PPY
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.