Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Pappalysin-1/PAPP-A Quantikine ELISA Kit
SDP

Catalog No. DPPA00 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DPPA00 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DPPA00 Supplier R&D Systems™ Supplier No. DPPA00
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Pappalysin-1/PAPP-A Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Pappalysin-1/PAPP-A Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Pappalysin-1/PAPP-A in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).

Specifications

Assay Range 0.78 to 50 ng/mL
Assay Sensitivity 0.116 ng/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (EDTA, Heparin), Plasma (Heparin), Serum, Serum
Specificity Natural and Recombinant Human PAPP-A
For Use With (Application) ELISA
Gene ID (Entrez) 5069
Gene Symbol PAPPA
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4 ase, IGFBP-4 aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA, PAPPA1, PAPP-A1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, Pappalysin1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1
Target Pappalysin-1/PAPP-A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.