Learn More
ACROBiosystems Human PAR2 500ug
Supplier:  ACROBiosystems PA2H52P7500UG
Encompass_Preferred
Human PAR2 Full Length Protein (VLP) (PA2-H52P7) is expressed from human 293 cells (HEK293). It contains AA Ser 37 - Tyr 397 (Accession # P55085).
Catalog No. 50-253-4333
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.