Learn More
ATCC Human parainfluenza virus 4b, strain CH 19503 (ATCC VR-1377)
Supplier:  ATCC VR1377
Encompass
Human parainfluenza virus 4b (strain CH 19503) is a respiratory virus isolate supplied by ATCC for infectious disease and respiratory disease research. The strain was deposited by the Centers for Disease Control and Prevention and is suitable for propagation in cell culture.
  • Isolated from a child with a respiratory tract infection in Washington, DC.
  • Strain CH 19503.
  • Propagated in NCI-H292 (H292) cell culture.
  • Stored frozen at -70°C or colder.
  • Intended for infectious disease and respiratory disease research.
  • Handle under appropriate biosafety conditions.
Catalog No. NC3834024