Learn More
ATCC Human parechovirus 1; Strain: Harris
Supplier:  ATCC VR52
Encompass
Human Parechovirus 2 strain Harris is propagated in monkey kidney cells. This strain was isolated in 1956 from the rectal swab of a child with diarrhea in Ohio. This product has applications in enteric disease research.
Catalog No. 50-238-2415