Learn More
Sino Biological Human PARP1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 310193MM06100
Encompass_Preferred
Produced in mouse immunized with E. coli-derived Human PARP / PARP1 fragment. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-0606
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.