Learn More
R&D Systems™ Human Patched 2/PTCH2 (NP_003729) VersaClone cDNA
SDP
Catalog No. RDC1059 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
RDC1059 10 μg
1 options
This item is not returnable. View return policy
Catalog No. RDC1059 Supplier R&D Systems™ Supplier No. RDC1059
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
ptc2; PTCH2
The Human Patched 2/PTCH2 (NP_003729) VersaClone cDNA clone contains the complete ORF for Patched 2/PTCH2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Patched 2/PTCH2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Line R&D Systems
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans