Learn More
R&D Systems™ Human Pax9 DuoSet ELISA Kit
Description
- Economical alternative to complete kits
- Development protocols are provided to guide further assay optimization
- Suggested diluent is suitable for the analysis of most cell culture supernate samples
Specifications
Specifications
| Assay Range | 46.90 to 3,000pg/mL |
| Conjugate | Biotin |
| Immunoassay Kit Format | 96-well Strip Plate |
| Product Type | Solid Phase Sandwich ELISA |
| Sufficient For | Five 96-well plates |
| For Use With (Application) | ELISA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.