Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PC-LKC Partial ORF (NP_060145, 210 a.a. - 318 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89956042 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-042 10 μg
89-956-043 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-956-042 Supplier Abnova™ Supplier No. H00054825Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the protocadherin family, which represents a subset of the larger cadherin superfamily. The members of the protocadherin family encode non-classical cadherins that function as calcium-dependent cell-cell adhesion molecules. The kidney-expressed gene product has a signal peptide, nine cadherin repeat domains and a unique cytoplasmic region. This protocadherin represents a new candidate for tumor suppression. [provided by RefSeq]

Sequence: GGMYHNTFTIQCSLPVFLSISVVDQPDLDPQFVREFYSASVAEDAAKGTSVLTVEAVDGDKGINDPVIYSISYSTRPGWFDIGADGVIRVNGSLDREQLLEADEEVQLQ

Specifications

Accession Number NP_060145
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 54825
Molecular Weight (g/mol) 37.73kDa
Name PC-LKC (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen GGMYHNTFTIQCSLPVFLSISVVDQPDLDPQFVREFYSASVAEDAAKGTSVLTVEAVDGDKGINDPVIYSISYSTRPGWFDIGADGVIRVNGSLDREQLLEADEEVQLQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ20124/FLJ20383/MGC163154/PC-LKC/PCLKC
Common Name PCDH24
Gene Symbol PCDH24
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.