Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PCDHAC2 Full-length ORF (NP_114089.1, 1 a.a. - 884 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89955909 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-955-909 10 μg
89-955-910 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-955-909 Supplier Abnova™ Supplier No. H00056134P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined. [provided by RefSeq]

Sequence: MEQAGTRPAATEHPRLRRPMPWLLLLPLLLLLLLLLPGPAASQLRYSVPEEQAPGALVGNVARALGLELRRLGPGCLRINHLGAPSPRYLELDLTSGALFVNERIDREALCEQRPRCLLSLEVLAHNPVAVSAVEVEILDINDNSPRFPRPNYQLQVSESVAPGARFHIESAQDPDVGANSVQTYELSPSEHFELDLKPLQENSKVLELVLRKGLDREQAALHHLVLTAVDGGIPARSGTAQISVRVLDTNDNSPAFDQSTYRVQLREDSPPGTLVVKLNASDPDEGSNGELRYSLSSYTSDRERQLFSIDASTGEVRVIGGLDYEEASSYQIYVQATDRGPVPMAGHCKVLVDIVDVNDNAPEVVLTDLYSPVPENATPNTIVAVLSVNDQDSGPNRKVSLGLEATLPFRLNGFGNSYTLVVSGPLDRERVAVYNITVTATDGGIPQLTSLRTLKVEISDINDNPPSFLEDSYSIYIQENNLPGVLLCTVQATDPDEKENAEVTYSLLEREIQGLPVTSYVSINSASGSLYAVNSFDYEKFREFFVTVEAQDKGSPPLSSTVTANVYVVDMNDHAPHILYPTSTNSSAAFEMVPRTAPAGYLVTKVIAMDSDSGQNAWLFYHLAQTSDLDLFKVELHTGEIRTTRKMGDESGSTFNLTVVVRDNGEPSLSASVAITVAVVDRVSKILPDTQRHVKSPRTYSEITLYLIIALSTVSFIFLLTIIILSIIKCYRYTAYGTACCGGFCGVRERSPAELYKQANNNIDARIPHGLKVQPHFIEVRGNGSLTKTYCYKACLTAGSGSDTFMFYNTGAQTGPGPSGAQAAVTDSRNLTGQSGQNAGNLIILKNEAVSQNEVRQWSGGLLQTHAFVTHPPISCDLALLSH

Specifications

Accession Number NP_114089.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 56134
Molecular Weight (g/mol) 122.6kDa
Name PCDHAC2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MEQAGTRPAATEHPRLRRPMPWLLLLPLLLLLLLLLPGPAASQLRYSVPEEQAPGALVGNVARALGLELRRLGPGCLRINHLGAPSPRYLELDLTSGALFVNERIDREALCEQRPRCLLSLEVLAHNPVAVSAVEVEILDINDNSPRFPRPNYQLQVSESVAPGARFHIESAQDPDVGANSVQTYELSPSEHFELDLKPLQENSKVLELVLRKGLDREQAALHHLVLTAVDGGIPARSGTAQISVRVLDTNDNSPAFDQSTYRVQLREDSPPGTLVVKLNASDPDEGSNGELRYSLSSYTSDRERQLFSIDASTGEVRVIGGLDYEEASSYQIYVQATDRGPVPMAGHCKVLVDIVDVNDNAPEVVLTDLYSPVPENATPNTIVAVLSVNDQDSGPNRKVSLGLEATLPFRLNGFGNSYTLVVSGPLDRERVAVYNITVTATDGGIPQLTSLRTLKVEISDINDNPPSFLEDSYSIYIQENNLPGVLLCTVQATDPDEKENAEVTYSLLEREIQGLPVTSYVSINSASGSLYAVNSFDYEKFREFFVTVEAQDKGSPPLSSTVTANVYVVDMNDHAPHILYPTSTNSSAAFEMVPRTAPAGYLVTKVIAMDSDSGQNAWLFYHLAQTSDLDLFKVELHTGEIRTTRKMGDESGSTFNLTVVVRDNGEPSLSASVAITVAVVDRVSKILPDTQRHVKSPRTYSEITLYLIIALSTVSFIFLLTIIILSIIKCYRYTAYGTACCGGFCGVRERSPAELYKQANNNIDARIPHGLKVQPHFIEVRGNGSLTKTYCYKACLTAGSGSDTFMFYNTGAQTGPGPSGAQAAVTDSRNLTGQSGQNAGNLIILKNEAVSQNEVRQWSGGLLQTHAFVTHPPISCDLALLSH
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC71598/PCDH-ALPHA-C2
Common Name PCDHAC2
Gene Symbol PCDHAC2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.