Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PCPE-1 ELISA Kit

Catalog No. EH362RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH362RB 96 Tests
EH362RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH362RB Supplier Invitrogen™ Supplier No. EH362RB
Add to Cart
Add to Cart

ELISA

Human PCPE-1 quantitates human PCPE-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PCPE-1.

Fibrillar collagen types I-III are synthesized as precursor molecules known as procollagens. These precursors contain amino- and carboxyl-terminal peptide extensions known as N- and C-propeptides, respectively, which are cleaved, upon secretion of procollagen from the cell, to yield the mature triple helical, highly structured fibrils. This gene encodes a glycoprotein which binds and drives the enzymatic cleavage of type I procollagen and heightens C-proteinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number astroglial cell transcript 2; Astt2; Astt-2; P14; Pcolce; PCPE; PCPE1; PCPE-1; procollagen C-endopeptidase enhancer; procollagen C-endopeptidase enhancer 1; procollagen C-endopeptidase enhancer protein; procollagen COOH-terminal proteinase enhancer 1; procollagen C-proteinase enhancer 1; procollagen C-proteinase enhancer protein; procollagen, type 1, COOH-terminal proteinase enhancer; Type 1 procollagen C-proteinase enhancer protein; Type I procollagen COOH-terminal proteinase enhancer
Assay Range 51 to 12,500 pg/mL
Assay Sensitivity 51 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5118
Gene Symbol PCOLCE
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target PCOLCE
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.