Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PCSK9 (NARC1) His-tag Recombinant Protein

Catalog No. PIRP88141
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88141 50 μg
PIRP88142 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP88141 Supplier Invitrogen™ Supplier No. RP88141
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (500 μL) be added to the vial to prepare a stock solution of 1.11 mg/mL. Concentration is measured by UV.

PCSK9 is a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. The protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NBP7
For Use With (Application) Control, ELISA Standard, Binding Assay
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 255738
Molecular Weight (g/mol) 72.5 kDa
Name Human PCSK9 (NARC1) His-tag
pH Range 7.4
Purification Method Purified
Quantity 50 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias AI415265; AI747682; convertase subtilisin; convertase subtilisin/kexin type 9 preproprotein; FH3; HCHOLA3; hypercholesterolemia; LDLCQ1; NARC1; NARC-1; neural apoptosis regulated convertase 1; neural apoptosis-regulated convertase 1; OTTHUMP00000009833; PC9; PCSK9; Proprotein convertase 9; proprotein convertase PC9; proprotein convertase subtilisin/kexin type 9; PSEC0052; Subtilisin/kexin-like protease PC9
Common Name PCSK9
Gene Symbol PCSK9
Biological Activity Loaded Recombinant Human LDLR/LDL Receptor Protein, mFc Tag on ProA Biosensor, can bind Recombinant Human PCSK9 Protein, His Tag with an affinity constant of 16 nM as determined in BLI assay. Immobilized Anti-PCSK9(Amg)-IgG2 Antibody (at 2 μg/mL (100 μL/well) can bind Recombinant Human PCSK9 / NARC1 Protein (His Tag), the EC50 is 14-52 ng/mL)
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence Human PCSK9, amino acids Met1-Gln692 (Accession # NP_777596.2) with a C-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Loaded Recombinant Human LDLR/LDL Receptor Protein, mFc Tag on ProA Biosensor, can bind Recombinant Human PCSK9 Protein, His Tag with an affinity constant of 16nM as determined in BLI assay. Immobilized Anti-PCSK9(Amg)-IgG2 Antibody at 2 μg/mL (100 μL/well) can bind Recombinant Human PCSK9 / NARC1 Protein (His Tag), the EC50 is 14-52 ng/mL.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.