Learn More
Description
- Also known as: FH3; PC9; NARC1; LDLCQ1; NARC-1; HCHOLA3
- PCSK9 is a member of the subtilisinlike proprotein convertase family, which includes proteases that process protein and peptide precursors trafficking through regulated or constitutive branches of the secretory pathway.
- It is expressed in liver, intestine and kidney tissues and escorts specific receptors for lysosomal degradation.
- It plays a role in cholesterol and fatty acid metabolism.
- Mutations in PCSK9 have been associated with autosomal dominant familial hypercholesterolemia.
- Alternatively spliced transcripts encoding different proteins have been described.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | Proprotein Convertase 9/PCSK9 |
| Accession Number | NP_777596 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2092 bp |
| Gene ID (Entrez) | 255738 |
| Gene Alias | EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1 convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
