Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PCTK1 Full-length ORF (NP_006192.1, 1 a.a. - 496 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89956106 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-106 10 ug
89-956-107 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-956-106 Supplier Abnova™ Supplier No. H00005127P02S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene belongs to the cdc2/cdkx subfamily of the ser/thr family of protein kinases. It may play a role in signal transduction cascades in terminally differentiated cells. This gene is thought to escape X inactivation. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: MDRMKKIKRQLSMTLRGGRGIDKTNGAPEQIGLDESGGGGGSDPGEAPTRAAPGELRSARGPLSSAPEIVHEDLKMGSDGESDQASATSSDEVQSPVRVRMRNHPPRKISTEDINKRLSLPADIRLPEGYLEKLTLNSPIFDKPLSRRLRRVSLSEIGFGKLETYIKLDKLGEGTYATVYKGKSKLTDNLVALKEIRLEHEEGAPCTAIREVSLLKDLKHANIVTLHDIIHTEKSLTLVFEYLDKDLKQYLDDCGNIINMHNVKLFLFQLLRGLAYCHRQKVLHRDLKPQNLLINERGELKLADFGLARAKSIPTKTYSNEVVTLWYRPPDILLGSTDYSTQIDMWGVGCIFYEMATGRPLFPGSTVEEQLHFIFRILGTPTEETWPGILSNEEFKTYNYPKYRAEALLSHAPRLDSDGADLLTKLLQFEGRNRISAEDAMKHPFFLSLGERIHKLPDTTSIFALKEIQLQKEASLRSSSMPDSGRPAFRVVDTEF

Specifications

Accession Number NP_006192.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5127
Molecular Weight (g/mol) 82.1kDa
Name PCTK1 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MDRMKKIKRQLSMTLRGGRGIDKTNGAPEQIGLDESGGGGGSDPGEAPTRAAPGELRSARGPLSSAPEIVHEDLKMGSDGESDQASATSSDEVQSPVRVRMRNHPPRKISTEDINKRLSLPADIRLPEGYLEKLTLNSPIFDKPLSRRLRRVSLSEIGFGKLETYIKLDKLGEGTYATVYKGKSKLTDNLVALKEIRLEHEEGAPCTAIREVSLLKDLKHANIVTLHDIIHTEKSLTLVFEYLDKDLKQYLDDCGNIINMHNVKLFLFQLLRGLAYCHRQKVLHRDLKPQNLLINERGELKLADFGLARAKSIPTKTYSNEVVTLWYRPPDILLGSTDYSTQIDMWGVGCIFYEMATGRPLFPGSTVEEQLHFIFRILGTPTEETWPGILSNEEFKTYNYPKYRAEALLSHAPRLDSDGADLLTKLLQFEGRNRISAEDAMKHPFFLSLGERIHKLPDTTSIFALKEIQLQKEASLRSSSMPDSGRPAFRVVDTEF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ16665/PCTAIRE/PCTAIRE1/PCTGAIRE
Common Name PCTK1
Gene Symbol PCTK1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.