Learn More
ACROBiosystems Human PD-1 25ug
Supplier:  ACROBiosystems PD1H528625UG
Encompass_Preferred
Human PD-1 Protein Twin-StrepTag (PD1-H5286) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # Q15116-1).
Catalog No. 50-253-4316
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.