Learn More
ACROBiosystems HEK293/Human PD-1 Stable Cell Line, Adherent, 2/Pk.
Catalog No. 502719754
Encompass_Preferred
The HEK293/Human PD-1 Stable Cell Line is engineered to express human PD-1 (Uniprot: Q15116). This adherent cell line is designed for use in cell-based PD-1 binding assays, with surface expression of human PD-1 confirmed by flow cytometry. Each vial contains at least 5×106 cells in 1 mL of serum-free cryopreservation medium.
  • Confirmed surface expression of human PD-1 via flow cytometry
  • Validated for use in cell-based binding assays
  • Maintained with Hygromycin B (20 μg/mL) selection marker
  • Requires complete growth medium of DMEM plus 10% FBS
  • Tested and verified negative for mycoplasma and sterility
  • Supplied as frozen cells in liquid nitrogen
Catalog No. 50-271-9754 Supplier ACROBiosystems Supplier No. CHEKATP1432VIALS
The HEK293/Human PD-1 Stable Cell Line is engineered to express human PD-1 (Uniprot: Q15116). This adherent cell line is designed for use in cell-based PD-1 binding assays, with surface expression of human PD-1 confirmed by flow cytometry. Each vial contains at least 5×106 cells in 1 mL of serum-free cryopreservation medium.
  • Confirmed surface expression of human PD-1 via flow cytometry
  • Validated for use in cell-based binding assays
  • Maintained with Hygromycin B (20 μg/mL) selection marker
  • Requires complete growth medium of DMEM plus 10% FBS
  • Tested and verified negative for mycoplasma and sterility
  • Supplied as frozen cells in liquid nitrogen