Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PD-1 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF1086SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1086SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1086SP Supplier R&D Systems Supplier No. AF1086SP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 26 publications

PD-1 Polyclonal specifically detects PD-1 in Cercocebus atys (Sooty Mangabey)∣Chinese Hamster∣Feline∣Human∣Mouse∣Primate∣Rhesus Macaque samples. It is validated for Bioassay∣Blockade of Receptor-ligand Interaction∣CyTOF∣Dual RNAScope ISH-IHC Compatible∣ELISA Capture (Matched Antibody Pair)∣ELISA Development∣Flow Cytometry∣Functional Assay∣Immunocytochemistry∣Immunohistochemistry∣Immunohistochemistry (Paraffin)∣Microarray∣Neutralization∣Surface Plasmon Resonance (SPR)∣Western Blot.

Specifications

Antigen PD-1
Applications Bioassay, Blockade of Receptor-ligand Interaction, CyTOF, Dual RNAScope ISH-IHC, ELISA Capture (Matched Antibody Pair), ELISA Development, Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Microarray, Neutralization, Surface Plasmon Resonance, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8IX89
Gene Alias CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Gene Symbols PDCD1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human PD-1 Leu25-Gln167 Accession # Q8IX89
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5133
Test Specificity Detects human PD-1 in ELISAs and Western blots. In sandwich ELISAs, less than 2% cross-reactivity with recombinant mouse PD-1 and less than 0.2% cross-reactivity with recombinant human (rh) CD28, rhICOS, and rhCTLA-4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Sooty Mangabey, Chinese Hamster, Feline, Human, Mouse, Primate, Rhesus Macaque
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.