Learn More
ACROBiosystems PD-L1 : PD-1 [Biotinylated] Inhibitor Screening ELISA Kit
Supplier:  ACROBiosystems EP13396TESTS
Encompass_Preferred
Immune checkpoint pathway is a focal point of today’s cancer research. PD-1 is one of the best characterized checkpoint proteins. The binding between PD-1 and its ligand PD-L1 suppresses T-cell activation and allows cancer cells to escape from body’s immune surveillance. Therefore, the pharmaceutical inhibition of PD-1 or its ligand has been considered a promising strategy by many oncologists.
Catalog No. 50-233-1639
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.