Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PDGF-B Recombinant Protein

Catalog No. PIRP75691
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP75691 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP75691 Supplier Invitrogen™ Supplier No. RP75691
Add to Cart
Add to Cart

Recombinant Protein

This product contains purified recombinant human PDGF B protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

This gene encodes a member of the protein family comprised of both platelet-derived growth factors (PDGF) and vascular endothelial growth factors (VEGF). The encoded preproprotein is proteolytically processed to generate platelet-derived growth factor subunit B, which can homodimerize, or alternatively, heterodimerize with the related platelet-derived growth factor subunit A. These proteins bind and activate PDGF receptor tyrosine kinases, which play a role in a wide range of developmental processes. Mutations in this gene are associated with meningioma. Reciprocal translocations between chromosomes 22 and 17, at sites where this gene and that for collagen type 1, alpha 1 are located, are associated with dermatofibrosarcoma protuberans, a rare skin tumor. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01127
Concentration 0.11 mg/mL
For Use With (Application) Control, Western Blot
Formulation 0.025 M sodium acetate with 1 mM EDTA, 50% glycerol and no preservative; pH 4.8
Gene ID (Entrez) 5155
Name Human PDGF-B
pH Range 4.8
Quantity 10 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias becaplermin; c-sis; I79_017626; IBGC5; PDGF; pdgf protein; PDGF subunit B; PDGF, B chain; PDGF2; PDGF-2; Pdgfb; PDGF-B; platelet derived growth factor subunit B; platelet derived growth factor, B polypeptide; platelet-derived growth factor 2; platelet-derived growth factor B; platelet-derived growth factor B chain; platelet-derived growth factor B subunit; platelet-derived growth factor beta; platelet-derived growth factor beta isoform 1, preproprotein; platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); Platelet-derived growth factor subunit B; platelet-derived growth factor, beta polypeptide (oncogene SIS); Platelet-derived growth factor, same as Sis (Simian sarcoma viral oncogene homologue, c-sis); proto-oncogene c-Sis; Sis; SSV; v-sis platelet-derived growth factor beta polypeptide (simian sarcoma viral oncogene homolog)
Common Name PDGF-B
Gene Symbol PDGFB
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Ser82-Thr190 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.