Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human PDGF-CC Recombinant Protein, PeproTech®

Catalog No. 10000CC5UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 250 μg
20 μg
250 μg
4 x 250 μg
5 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10000CC5UG 5 μg
10-000-CC01M 4 x 250 μg
10000CC20UG 20 μg
50-272-2982 2 x 250 μg
10000CC250 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 10000CC5UG Supplier Gibco™ Supplier No. 10000CC5UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

100-00CC-1MG will be provided as 4 x 250 μg (100-00CC-250UG). 100-00CC-500UG will be provided as 2 x 250 μg (100-00CC-250UG). Recombinant Human PDGF-CC is a 25.0 kDa protein consisting of two identical disulfide-linked, 112 amino acid, polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The platelet-derived growth factor (PDGF) family of heparin-binding growth factors consists of five known members, denoted PDGF-AA, PDGF-BB, PDGF-AB, PDGF-CC and PDGF-DD. The mature and active form of these proteins, an anti-parallel, disulfide-linked dimer of two 12-14 kDa, polypeptide chains, is obtained through proteolytic processing of biologically inactive precursor proteins, which contain an N-terminal CUB domain and a PDGF/VEGF homologous domain. The PDGFs interact with two related protein tyrosine kinase receptors, PDGFR-alpha and PDGFR-beta, and are potent mitogens for a variety of cell types, including smooth muscle cells, connective tissue cells, bone and cartilage cells, and certain tumor cells. They play an important role in a number of biological processes, including hyperplasia, chemotaxis, embryonic neuron development, and respiratory tubules' epithelial cell development. Mature PDGFs are stored in platelet alpha-granules, and are released upon platelet activation. PDGF-AA, -AB, -BB and -CC signal primarily through the PDGFR-alpha receptor, whereas PDGF-DD interacts almost exclusively with the PDGFR-beta receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NRA1
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 56034
Molecular Weight (g/mol) 25 kDa
Name Human PDGF-CC
Purification Method Purified
Quantity 5 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias PDGF-C; Platelet Derived Growth Factor C; SCDGF; Spinal Cord-Derived Growth Factor
Common Name PDGF-CC
Gene Symbol PDGFC
Biological Activity Determined by the dose-dependent stimulation of the proliferation of Balb/c 3T3 cells. The expected The ED50 for this effect is 15-20 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MVVDLNLLTE EVRLYSCTPR NFSVSIREEL KRTDTIFWPG CLLVKRCGGN CACCLHNCNE CQCVPSKVTK KYHEVLQLRP KTGVRGLHKS LTDVALEHHE ECDCVCRGST GG
Content And Storage -20°C
Activity Determined by the dose-dependent stimulation of the proliferation of Balb/c 3T3 cells. The expected The ED50 for this effect is 15-20 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.