Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PDLIM7 Full-length ORF (AAH01093.1, 1 a.a. - 457 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89956333 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-333 25 μg
89-956-332 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-956-333 Supplier Abnova™ Supplier No. H00009260P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is representative of a family of proteins composed of conserved PDZ and LIM domains. LIM domains are proposed to function in protein-protein recognition in a variety of contexts including gene transcription and development and in cytoskeletal interaction. The LIM domains of this protein bind to protein kinases, whereas the PDZ domain binds to actin filaments. The gene product is involved in the assembly of an actin filament-associated complex essential for transmission of ret/ptc2 mitogenic signaling. The biological function is likely to be that of an adapter, with the PDZ domain localizing the LIM-binding proteins to actin filaments of both skeletal muscle and nonmuscle tissues. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq]

Sequence: MDSFKVVLEGPAPWGFRLQGGKDFNVPLSISRLTPGGKAAQAGVAVGDWVLSIDGENAGSLTHIEAQNKIRACGERLSLGLSRAQPVQSKPQKASAPAADPPRYTFAPSVSLNKTARPFGAPPPADSAPQQNGQPLRPLVPDASKQRLMENTEDWRPRPGTGQSRSFRILAHLTGTEFMQDPDEEHLKKSSQVPRTEAPAPASSTPQEPWPGPTAPSPTSRPPWAVDPAFAERYAPDKTSTVLTRHSQPATPTPLQSRTSIVQAAAGGVPGGGSNNGKTPVCHQCHKVIRGRYLVALGHAYHPEEFVCSQCGKVLEEGGFFEEKGAIFCPPCYDVRYAPSCAKCKKKITGEIMHALKMTWHVHCFTCAACKTPIRNRAFYMEEGVPYCERDYEKMFGTKCHGCDFKIDAGDRFLEALGFSWHDTCFVCAICQINLEGKTFYSKKDRPLCKSHAFSHV

Specifications

Accession Number AAH01093.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9260
Molecular Weight (g/mol) 76.01kDa
Name PDLIM7 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MDSFKVVLEGPAPWGFRLQGGKDFNVPLSISRLTPGGKAAQAGVAVGDWVLSIDGENAGSLTHIEAQNKIRACGERLSLGLSRAQPVQSKPQKASAPAADPPRYTFAPSVSLNKTARPFGAPPPADSAPQQNGQPLRPLVPDASKQRLMENTEDWRPRPGTGQSRSFRILAHLTGTEFMQDPDEEHLKKSSQVPRTEAPAPASSTPQEPWPGPTAPSPTSRPPWAVDPAFAERYAPDKTSTVLTRHSQPATPTPLQSRTSIVQAAAGGVPGGGSNNGKTPVCHQCHKVIRGRYLVALGHAYHPEEFVCSQCGKVLEEGGFFEEKGAIFCPPCYDVRYAPSCAKCKKKITGEIMHALKMTWHVHCFTCAACKTPIRNRAFYMEEGVPYCERDYEKMFGTKCHGCDFKIDAGDRFLEALGFSWHDTCFVCAICQINLEGKTFYSKKDRPLCKSHAFSHV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias LMP1
Common Name PDLIM7
Gene Symbol PDLIM7
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.