Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PEAR1 ELISA Kit

Catalog No. EH366RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH366RBX10 10 x 96 Tests
EH366RBX5 5 x 96 Tests
EH366RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH366RBX10 Supplier Invitrogen™ Supplier No. EH366RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Human PEAR1 quantitates human PEAR1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PEAR1.

Platelet endothelial aggregation receptor 1 (PEAR1) is a platelet receptor that signals upon the formation of platelet-platelet contacts independent of platelet activation and secondary to platelet aggregation. Upon platelet aggregation stimulated by physiological agonists, PEAR1 becomes tyrosine- and serine-phosphorylated; the tyrosine phosphorylation can be inhibited by eptifibatide, an aIIbbeta3 antagonist that also inhibits platelet aggregation. Recent studies have indicated that genetic variations in PEAR1, may be associated with enhanced agonist-induced platelet aggregation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 3110045G13Rik; hPEAR1; Jagged and Delta protein; Jedi; Jedi-1; Megf12; mPEAR1; multiple EGF-like domains protein 12; multiple EGF-like-domains 12; multiple epidermal growth factor-like domains protein 12; Pear1; platelet endothelial aggregation receptor 1; Protein Jedi; RGD1305653; RP11-107D16.3
Assay Range 0.45 to 100 ng/mL
Assay Sensitivity 0.45 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 375033
Gene Symbol PEAR1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target PEAR1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.