Learn More
Description
Specifications
Specifications
| Assay Range | 0.31 to 20 ng/mL |
| Assay Sensitivity | 0.026 ng/mL |
| Sample Type | Plasma (EDTA), Saliva, Serum-free Cell Culture Supernatant |
| Specificity | Natural and Recombinant Human Pentraxin 3 |
| Gene ID (Entrez) | 5806 |
| Host Species | Human |
| Quantity | 1 Kit |
| Sample Volume | Serum-free Cell Culture Supernates (20 μL),EDTA Plasma (20 μL),Saliva (20 μL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | alpha-induced protein 5, pentaxin-related gene, rapidly induced by IL-1 beta, tumor necrosis factor, Pentaxin-related protein PT x 3, pentraxin 3, long, pentraxin-3, pentraxin-related gene, rapidly induced by IL-1 beta, pentraxin-related protein PT x 3, PT x 3, TNF alpha-induced protein 5, TNFAIP5, TSG14, TSG-14, TSG-14 pentaxin-related gene, rapidly induced by IL-1 beta, Tumor necrosis factor alpha-induced protein 5, tumor necrosis factor, alpha-induced protein 5, Tumor necrosis factor-inducible gene 14 protein, tumor necrosis factor-inducible protein TSG-14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
