Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PER3 Full-length ORF (AAH26102.1, 1 a.a. - 378 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89956433 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-433 25 μg
89-956-432 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-956-433 Supplier Abnova™ Supplier No. H00008863P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the Period family of genes and is expressed in a circadian pattern in the suprachiasmatic nucleus, the primary circadian pacemaker in the mammalian brain. Genes in this family encode components of the circadian rhythms of locomotor activity, metabolism, and behavior. Circadian expression in the suprachiasmatic nucleus continues in constant darkness, and a shift in the light/dark cycle evokes a proportional shift of gene expression in the suprachiasmatic nucleus. The specific function of this gene is not yet known. [provided by RefSeq]

Sequence: MPRGEAPGPGRRGAKDEALGEESGERWSPEFHLQRKLADSSHSEQQDRNRVSEELIMVVQEMKKYFPSERRNKPSTLDALNYALRCVHSVQANSEFFQILSQNGAPQADVSMYSLEELATIASEHTSKNTDTFVAVFSFLSGRLVHISEQAALILNRKKDVLASSHFVDLLAPQDMRVFYAHTARAQLPFWNNWTQRAARYECAPVKPFFCRIRGGEDRKQEKCHSPFRIIPYLIHVHHPAQPELESEPCCLTVVEKIHSGYEAPRIPVNKRIFTTTHTPGCVFLEVDEKAVPLLGYLPQDLIGTSILSYLHPEDRSLMVAIHQKVLKYAGHPPFEHSPIRFCTQNGDYIILDSSWSSFVNPWSRKISFIIGRHKVRT

Specifications

Accession Number AAH26102.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8863
Molecular Weight (g/mol) 69.5kDa
Name PER3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MPRGEAPGPGRRGAKDEALGEESGERWSPEFHLQRKLADSSHSEQQDRNRVSEELIMVVQEMKKYFPSERRNKPSTLDALNYALRCVHSVQANSEFFQILSQNGAPQADVSMYSLEELATIASEHTSKNTDTFVAVFSFLSGRLVHISEQAALILNRKKDVLASSHFVDLLAPQDMRVFYAHTARAQLPFWNNWTQRAARYECAPVKPFFCRIRGGEDRKQEKCHSPFRIIPYLIHVHHPAQPELESEPCCLTVVEKIHSGYEAPRIPVNKRIFTTTHTPGCVFLEVDEKAVPLLGYLPQDLIGTSILSYLHPEDRSLMVAIHQKVLKYAGHPPFEHSPIRFCTQNGDYIILDSSWSSFVNPWSRKISFIIGRHKVRT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GIG13
Common Name PER3
Gene Symbol PER3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.