Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Perforin QuicKit ELISA
SDP

Catalog No. SK8011 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Pack (6 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SK8011 1 Pack (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SK8011 Supplier R&D Systems™ Supplier No. SK8011
Add to Cart
Add to Cart

80-minute solid phase ELISA designed to measure human Perforin levels in cell culture supernates, serum, plasma, and cell lysates

Specifications

Specificity Natural and recombinant human Perforin
Target Species Human
Gene ID (Entrez) 5551
Quantity 1 Pack (6 Plates)
Storage Requirements Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Synonym Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFP, PFPcytolysin, PRF1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.