Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Persephin Recombinant Protein, PeproTech®

Catalog No. 45012100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45012100UG 100 μg
450-12-1MG 1 mg
450-12-20UG 20 μg
45012250UG 250 μg
45012500UG 500 μg
450-12-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 45012100UG Supplier Gibco™ Supplier No. 45012100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Persephin is a disulfide-linked homodimer, composed of two 10.4 kDa polypeptide chains (194 total amino acid residues). Each chain contains seven conserved cysteine residues, one of which (Cys 64) is used for inter-chain disulfide bridging, and the others are involved in the intramolecular ring formation known as the cysteine knot configuration. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Persephin is a disulfide-linked, homodimeric, neurotrophic factor structurally related to GDNF, artemin, and neurturin. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. Persephin signals through a multicomponent receptor system, composed of RET and one of four GFR alpha (alpha1-alpha4) receptors. The GFRalpha4 was first identified in chicken, and was later shown to be the preferential binding subunit for persephin. Persephin promotes the survival of ventral midbrain dopaminergic neurons and motor neurons after sciatic nerve oxotomy, and, like GDNF, promotes ureteric bud branching. However, in contrast to GDNF and neurturin, persephin does not support the survival of peripheral neurons.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60542
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 5623
Molecular Weight (g/mol) 20.8 kDa
Name Human Persephin
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Persephin; PSP; Pspn
Common Name PSPN
Gene Symbol PSPN
Biological Activity Human Persephin induces RET phosphorylation using a concentration range of 0.1-1.0 ng/ml Human Persephin binds to mammalian GFRa4 with the Kd of 100 pM. Other members of the GDNF family (Artemin, GDNF and Neurturin) do not bind to mammalian GFRa4.
Conjugate Unconjugated
Recombinant Recombinant
Sequence RALSGPCQLW SLTLSVAELG LGYASEEKVI FRYCAGSCPR GARTQHGLAL ARLQGQGRAH GGPCCRPTRY TDVAFLDDRH RWQRLPQLSA AACGCGG
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.