Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PEX6 Full-length ORF (AAH48331.1, 1 a.a. - 980 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89956483 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-483 25 ug
89-956-482 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-956-483 Supplier Abnova™ Supplier No. H00005190P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the AAA (ATPases associated with diverse cellular activities) family of ATPases. This member is a predominantly cytoplasmic protein, which plays a direct role in peroxisomal protein import and is required for PTS1 (peroxisomal targeting signal 1, a C-terminal tripeptide of the sequence ser-lys-leu) receptor activity. Mutations in this gene cause peroxisome biogenesis disorders of complementation group 4 and complementation group 6. [provided by RefSeq]

Sequence: MALAVLRVLEPFPTETPPLAVLLPPGGPWPAAELGLVLALRPAGESPAGPALLVAALEGPDAGTEEQGPGPPQLLVSRALLRLLALGSGAWVRARAVRRPPALGWALLGTSLGPGLGPRVGPLLVRRGETLPVPGPRVLETRPALQGLLGPGTRLAVTELRGRARLCPESGDSSRPPPPPVVSSFAVSGTVRRLQGVLGGTGDSLGVSRSCLRGLGLFQGEWVWVAQARESSNTSQPHLARVQVLEPRWDLSDRLGPGSGPLGEPLADGLALVPATLAFNLGCDPLEMGELRIQRYLEGSIAPEDKGSCSLLPGPPFARELHIEIVSSPHYSTNGNYDGVLYRHFQIPRVVQEGDVLCVPTIGQVEILEGSPEKLPRWREMFFKVKKTVGEAPDGPASAYLADTTHTSLYMVGSTLSPVPWLPSEESTLWSSLSPPGLEALVSELCAVLKPRLQPGGALLTGTSSVLLRGPPGCGKTTVVAAACSHLGLHLLKVPCSSLCAESSGAVETKLQAIFSRARRCRPAVLLLTAVDLLGRDRDGLGEDARVMAVLRHLLLNEDPLNSCPPLMVVATTSRAQDLPADVQTAFPHELEVPALSEGQRLSILRALTAHLPLGQEVNLAQLARRCAGFVVGDLYALLTHSSRAACTRIKNSGLAGGLTEEDEGELCAAGFPLLAEDFGQALEQLQTAHSQAVGAPKIPSVSWHDVGGLQEVKKEILETIQLPLEHPELLSLGLRRSGLLLHGPPGTGKTLLAKAVATECSLTFLSVKGPELINMYVGQSEENVREVFARARAAAPCIIFFDELDSLAPSRGRSGDSGGVMDRVVSQLLAELDGLHSTQDVFVIGATNRPDLLDPALLRPGRFDKLVFVGANEDRASQLRVLSAITRKFKLEPSVSLVNVLDCCPPQLTGADLYSLCSDAMTAALKRRVHDLEEGLEPGSSALMLTMEDLLQAAARLQPSVSEQELLRYKRIQRKFAAC

Specifications

Accession Number AAH48331.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5190
Molecular Weight (g/mol) 130.5kDa
Name PEX6 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MALAVLRVLEPFPTETPPLAVLLPPGGPWPAAELGLVLALRPAGESPAGPALLVAALEGPDAGTEEQGPGPPQLLVSRALLRLLALGSGAWVRARAVRRPPALGWALLGTSLGPGLGPRVGPLLVRRGETLPVPGPRVLETRPALQGLLGPGTRLAVTELRGRARLCPESGDSSRPPPPPVVSSFAVSGTVRRLQGVLGGTGDSLGVSRSCLRGLGLFQGEWVWVAQARESSNTSQPHLARVQVLEPRWDLSDRLGPGSGPLGEPLADGLALVPATLAFNLGCDPLEMGELRIQRYLEGSIAPEDKGSCSLLPGPPFARELHIEIVSSPHYSTNGNYDGVLYRHFQIPRVVQEGDVLCVPTIGQVEILEGSPEKLPRWREMFFKVKKTVGEAPDGPASAYLADTTHTSLYMVGSTLSPVPWLPSEESTLWSSLSPPGLEALVSELCAVLKPRLQPGGALLTGTSSVLLRGPPGCGKTTVVAAACSHLGLHLLKVPCSSLCAESSGAVETKLQAIFSRARRCRPAVLLLTAVDLLGRDRDGLGEDARVMAVLRHLLLNEDPLNSCPPLMVVATTSRAQDLPADVQTAFPHELEVPALSEGQRLSILRALTAHLPLGQEVNLAQLARRCAGFVVGDLYALLTHSSRAACTRIKNSGLAGGLTEEDEGELCAAGFPLLAEDFGQALEQLQTAHSQAVGAPKIPSVSWHDVGGLQEVKKEILETIQLPLEHPELLSLGLRRSGLLLHGPPGTGKTLLAKAVATECSLTFLSVKGPELINMYVGQSEENVREVFARARAAAPCIIFFDELDSLAPSRGRSGDSGGVMDRVVSQLLAELDGLHSTQDVFVIGATNRPDLLDPALLRPGRFDKLVFVGANEDRASQLRVLSAITRKFKLEPSVSLVNVLDCCPPQLTGADLYSLCSDAMTAALKRRVHDLEEGLEPGSSALMLTMEDLLQAAARLQPSVSEQELLRYKRIQRKFAAC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PAF-2/PAF2/PXAAA1
Common Name PEX6
Gene Symbol PEX6
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.