Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PHKA1 Partial ORF (NP_002628, 631 a.a. - 730 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89956704 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-704 10 ug
89-956-705 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-956-704 Supplier Abnova™ Supplier No. H00005255Q01S

Used for AP, Array, ELISA, WB-Re

The PHKA1 gene encodes the alpha subunit of muscle phosphorylase kinase (EC 2.7.1.38), a key regulatory enzyme of glycogen metabolism. Phosphorylase kinase consists of 4 copies of an alpha-beta-gamma-delta tetramer. The alpha, beta (PHKB; MIM 172490), and gamma (PHKG1; MIM 172470 and PHKG2; MIM 172471) subunits have several isoforms; the delta subunit is calmodulin (CALM1; MIM 114180). PHKA2 (MIM 306000) encodes the alpha subunit of liver-specific phosphorylase kinase and is also located on the X chromosome.[supplied by OMIM]

Sequence: DYDDNYDYLESGNWMNDYDSTSHARCGDEVARYLDHLLAHTAPHPKLAPTSQKGGLDRFQAAVQTTCDLMSLVTKAKELHVQNVHMYLPTKLFQASRPSF

Specifications

Accession Number NP_002628
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5255
Molecular Weight (g/mol) 36.74kDa
Name PHKA1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen DYDDNYDYLESGNWMNDYDSTSHARCGDEVARYLDHLLAHTAPHPKLAPTSQKGGLDRFQAAVQTTCDLMSLVTKAKELHVQNVHMYLPTKLFQASRPSF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC132604/PHKA
Common Name PHKA1
Gene Symbol PHKA1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.