Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-Chk2 (T68) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1626SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1626SP 25 μg
MAB1626100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1626SP Supplier R&D Systems Supplier No. MAB1626SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Chk2 Monoclonal specifically detects Chk2 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Chk2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1238H
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 2-25 ug/mL
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. O96017
Gene Alias CDS1, CHEK2, CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, HuCds1, LFS2, PP1425, Rad53, S.pombe) homolog
Gene Symbols CHEK2
Host Species Rabbit
Immunogen E.coli-derived recombinant human Phospho-Chk2 (T68) Accession # O96017
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11200
Test Specificity Detects human Chk2 when phosphorylated at T68 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.