Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-ErbB2/Her2 (Y1248) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1768SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1768SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1768SP Supplier R&D Systems Supplier No. AF1768SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 2 publications

ErbB2/Her2 Polyclonal specifically detects ErbB2/Her2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen ErbB2/Her2 (p Tyr1248)
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 0.3-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, Neu Oncogene, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGL, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, TKR1, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian)
Gene Symbols ERBB2
Host Species Rabbit
Immunogen Phosphopeptide containing human ErbB2 Y1248 site
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2064
Test Specificity Detects human ErbB2 when phosphorylated at Y1248.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.