Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Phospho-Flt-3/Flk-2 DuoSet IC ELISA

Catalog No. p-4754395
Change view
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DYC368E 15 Plates
DYC3682 2 Plates
DYC3685 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC368E Supplier R&D Systems™ Supplier No. DYC368E
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. This kit is designed for the detection and enumeration of IFN-γ/IL-2. Complete ELISpot kits are ready-to-run and require no assay development or refinement.

The Human Phospho-Flt-3/Flk-2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human Flt-3/Flk-2/CD135.

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit
Species Human
Conjugate Biotin
For Use With (Application) ELISA
Gene ID (Entrez) 2322
Packaging Quantity 15 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.