Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-Histone H2AX (S139) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2288SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2288SP 25 μg
MAB2288100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2288SP Supplier R&D Systems Supplier No. MAB2288SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Histone H2AX Monoclonal specifically detects Histone H2AX in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen Histone H2AX
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2207D
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P16104
Gene Alias H2A.X, H2A/X, H2AFX
Host Species Rabbit
Immunogen Phospho-peptide containing human Histone H2AX S139 site Accession # P16104
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 3014
Test Specificity Detects human Histone H2AX when phosphorylated at S139 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.