Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-Nbs1 (S343) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4944SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4944SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4944SP Supplier R&D Systems Supplier No. AF4944SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Nbs1 Polyclonal specifically detects Nbs1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Nbs1 (p ser343)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and Sodium Azide with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Alias ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBN, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex
Gene Symbols NBN
Host Species Rabbit
Immunogen Phosphopeptide containing human Nbs1 S343 site
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4683
Test Specificity Detects human Nbs1 when phosphorylated at S343. Does not recognize Nbs1 when unphosphorylated at S343.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.