Learn More
R&D Systems™ Human Phospho-p70 S6 Kinase (T389) DuoSet IC ELISA
Description
1 Kit (for 5 Plates)
Specifications
Specifications
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 6198 |
| Host Species | Human |
| Quantity | 1 Kit (2 Plates) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Target | p70 S6 Kinase/S6K |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.