Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-PRAS40 (T246) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB68901SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB68901SP 25 μg
MAB68901 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB68901SP Supplier R&D Systems Supplier No. MAB68901SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PRAS40 Monoclonal specifically detects PRAS40 in Human samples. It is validated for Western Blot.

Specifications

Antigen PRAS40
Applications Western Blot
Classification Monoclonal
Clone 1211A
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. Q96B36
Gene Alias 40 kDa, AKT1 substrate 1 (proline-rich), AKT1S1, Lobe, MGC2865, proline-rich AKT1 substrate 1,40 kDa proline-rich AKT substrate
Gene Symbols AKT1S1
Host Species Rabbit
Immunogen Phosphopeptide containing the human PRAS40 T246 site Accession # Q96B36
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84335
Test Specificity Detects human and mouse PRAS40 when phosphorylated at T246.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.