Learn More
Abnova™ Human PI15 Full-length ORF (NP_056970.1, 1 a.a. - 258 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
This gene encodes a trypsin inhibitor. The protein shares similarity to insect venom allergens, mammalian testis-specific proteins and plant pathogenesis-related proteins. It is frequently expressed in human neuroblastoma and glioblastoma cell lines, and thus may play a role in the central nervous system. [provided by RefSeq]
Specifications
Specifications
| Accession Number | NP_056970.1 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 51050 |
| Molecular Weight (g/mol) | 55.5kDa |
| Name | PI15 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 10 μg |
| Immunogen | MIAISAVSSALLFSLLCEASTVVLLNSTDSSPPTNNFTDIEAALKAQLDSADIPKARRKRYISQNDMIAILDYHNQVRGKVFPPAANMEYMVWDENLAKSAEAWAATCIWDHGPSYLLRFLGQNLSVRTGRYRSILQLVKPWYDEVKDYAFPYPQDCNPRCPMRCFGPMCTHYTQMVWATSNRIGCAIHTCQNMNVWGSVWRRAVYLVCNYAPKGNWIGEAPYKVGVPCSSCPPSYGGSCTDNLCFPGVTSNYLYWFK |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.