Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PI4KA Full-length ORF (NP_002641.1, 1 a.a. - 854 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89956975 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-975 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-956-975 Supplier Abnova™ Supplier No. H00005297P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a phosphatidylinositol (PI) 4-kinase which catalyzes the first committed step in the biosynthesis of phosphatidylinositol 4,5-bisphosphate. The mammalian PI 4-kinases have been classified into two types, II and III, based on their molecular mass, and modulation by detergent and adenosine. The protein encoded by this gene is a type III enzyme that is not inhibited by adenosine. Two transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Sequence: MREMAGAWHMTVEQKFGLFSAEIKEADPLAASEASQPKPCPPEVTPHYIWIDFLVQRFEIAKYCSSDQVEIFSSLLQRSMSLNIGGAKGSMNRHVAAIGPRFKLLTLGLSLLHADVVPNATIRNVLREKIYSTAFDYFSCPPKFPTQGEKRLREDISIMIKFWTAMFSDKKYLTASQLVPPDNQDTRSNLDITVGSRQQATQGWINTYPLSSGMSTISKKSGMSKKTNRGSQLHKYYMKRRTLLLSLLATEIERLITWYNPLSAPELELDQAGENSVANWRSKYISLSEKQWKDNVNLAWSISPYLAVQLPARFKNTEAIGNEVTRLVRLDPGAVSDVPEAIKFLVTWHTIDADAPELSHVLCWAPTDPPTGLSYFSSMYPPHPLTAQYGVKVLRSFPPDAILFYIPQIVQALRYDKMGYVREYILWAASKSQLLAHQFIWNMKTNIYLDEEGHQKDPDIGDLLDQLVEEITGSLSGPAKDFYQREFDFFNKITNVSAIIKPYPKGDERKKACLSALSEVKVQPGCYLPSNPEAIVLDIDYKSGTPMQSAAKAPYLAKFKVKRCGVSELEKEGLRCRSDSEDECSTQEADGQKISWQAAIFKVGDDCRQDMLALQIIDLFKNIFQLVGLDLFVFPYRVVATAPGCGVIECIPDCTSRDQLGRQTDFGMYDYFTRQYGDESTLAFQQARYNFIRSMAAYSLLLFLLQIKDRHNGNIMLDKKGHIIHIDFGFMFESSPGGNLGWEPDIKLTDEMVMIMGGKMEATPFKWFMEMCVRGYLAVRPYMDAVVSLVTLMLDTGLPCFRGQTIKLLKHRFSPNMTEREAANFIMKVIQSCFLSNRSRTYDMIQYYQNDIPY

Specifications

Accession Number NP_002641.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5297
Molecular Weight (g/mol) 123.4kDa
Name PI4KA (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MREMAGAWHMTVEQKFGLFSAEIKEADPLAASEASQPKPCPPEVTPHYIWIDFLVQRFEIAKYCSSDQVEIFSSLLQRSMSLNIGGAKGSMNRHVAAIGPRFKLLTLGLSLLHADVVPNATIRNVLREKIYSTAFDYFSCPPKFPTQGEKRLREDISIMIKFWTAMFSDKKYLTASQLVPPDNQDTRSNLDITVGSRQQATQGWINTYPLSSGMSTISKKSGMSKKTNRGSQLHKYYMKRRTLLLSLLATEIERLITWYNPLSAPELELDQAGENSVANWRSKYISLSEKQWKDNVNLAWSISPYLAVQLPARFKNTEAIGNEVTRLVRLDPGAVSDVPEAIKFLVTWHTIDADAPELSHVLCWAPTDPPTGLSYFSSMYPPHPLTAQYGVKVLRSFPPDAILFYIPQIVQALRYDKMGYVREYILWAASKSQLLAHQFIWNMKTNIYLDEEGHQKDPDIGDLLDQLVEEITGSLSGPAKDFYQREFDFFNKITNVSAIIKPYPKGDERKKACLSALSEVKVQPGCYLPSNPEAIVLDIDYKSGTPMQSAAKAPYLAKFKVKRCGVSELEKEGLRCRSDSEDECSTQEADGQKISWQAAIFKVGDDCRQDMLALQIIDLFKNIFQLVGLDLFVFPYRVVATAPGCGVIECIPDCTSRDQLGRQTDFGMYDYFTRQYGDESTLAFQQARYNFIRSMAAYSLLLFLLQIKDRHNGNIMLDKKGHIIHIDFGFMFESSPGGNLGWEPDIKLTDEMVMIMGGKMEATPFKWFMEMCVRGYLAVRPYMDAVVSLVTLMLDTGLPCFRGQTIKLLKHRFSPNMTEREAANFIMKVIQSCFLSNRSRTYDMIQYYQNDIPY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ16556/PI4K-ALPHA/PIK4CA/pi4K230
Common Name PI4KA
Gene Symbol PI4KA
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.