Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PIGC Full-length ORF (NP_002633.1, 1 a.a. - 297 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89956902 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-902 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-956-902 Supplier Abnova™ Supplier No. H00005279P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes an endoplasmic reticulum associated protein that is involved in glycosylphosphatidylinositol (GPI) lipid anchor biosynthesis. The GPI lipid anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. The encoded protein is one subunit of the GPI N-acetylglucosaminyl (GlcNAc) transferase that transfers GlcNAc to phosphatidylinositol (PI) on the cytoplasmic side of the endoplasmic reticulum. Two alternatively spliced transcripts that encode the same protein have been found for this gene. A pseudogene on chromosome 11 has also been characterized. [provided by RefSeq]

Sequence: MYAQPVTNTKEVKWQKVLYERQPFPDNYVDRRFLEELRKNIHARKYQYWAVVFESSVVIQQLCSVCVFVVIWWYMDEGLLAPHWLLGTGLASSLIGYVLFDLIDGGEGRKKSGQTRWADLKSALVFITFTYGFSPVLKTLTESVSTDTIYAMSVFMLLGHLIFFDYGANAAIVSSTLSLNMAIFASVCLASRLPRSLHAFIMVTFAIQIFALWPMLQKKLKACTPRSYVGVTLLFAFSAVGGLLSISAVGAVLFALLLMSISCLCPFYLIRLQLFKENIHGPWDEAEIKEDLSRFLS

Specifications

Accession Number NP_002633.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5279
Molecular Weight (g/mol) 60kDa
Name PIGC (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MYAQPVTNTKEVKWQKVLYERQPFPDNYVDRRFLEELRKNIHARKYQYWAVVFESSVVIQQLCSVCVFVVIWWYMDEGLLAPHWLLGTGLASSLIGYVLFDLIDGGEGRKKSGQTRWADLKSALVFITFTYGFSPVLKTLTESVSTDTIYAMSVFMLLGHLIFFDYGANAAIVSSTLSLNMAIFASVCLASRLPRSLHAFIMVTFAIQIFALWPMLQKKLKACTPRSYVGVTLLFAFSAVGGLLSISAVGAVLFALLLMSISCLCPFYLIRLQLFKENIHGPWDEAEIKEDLSRFLS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GPI2/MGC2049
Common Name PIGC
Gene Symbol PIGC
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.